Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,6'-((3,3'-Di-tert-butyl-5,5'-dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(oxy))didibenzo[d,f][1,3,2]dioxaphosphepine 98%

Supplier: AMBEED, INC
Warning

A169395-100MG A169395-1G A169395-250MG
77163-476EA 32 USD InStock
77163-476 77163-478 77163-480
6,6'-((3,3'-Di-tert-butyl-5,5'-dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(oxy))didibenzo[d,f][1,3,2]dioxaphosphepine 98%
6,6'-((3,3'-Di-tert-butyl-5,5'-dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(oxy))didibenzo[d,f][1,3,2]dioxaphosphepine
Precaución: for Research Use Only


Formula: C₄₆H₄₄O₈P₂
MDL Number: MFCD00233876
CAS Number: 121627-17-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty