Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide 95%

Supplier: AMBEED, INC
Warning

A169997-100MG A169997-10G A169997-1G A169997-250MG A169997-5G
77163-504EA 67 USD InStock
77163-504 77163-506 77163-508 77163-510 77163-512
2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide 95%
2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Precaución: for Research Use Only


Formula: C₆H₁₄Br₂N₂
MW: 274.00 g/mol
MDL Number: MFCD28134575
CAS Number: 114086-15-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty