Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrachloro-o-benzoquinone 97%

Supplier: AMBEED, INC
Warning

A171921-1G A171921-5G
77163-592EA 36 USD InStock
77163-592 77163-594
Tetrachloro-o-benzoquinone 97%
Tetrachloro-o-benzoquinone
Precaución: for Research Use Only


Formula: C₆Cl₄O₂
MDL Number: MFCD00001646
CAS Number: 2435-53-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty