Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-5-chloro-2-methoxyaniline 95%

Supplier: AMBEED, INC
Warning

A172385-10G A172385-1G A172385-250MG A172385-25G A172385-5G
77163-602EA 75 USD InStock
77163-602 77163-604 77163-606 77163-608 77163-610
4-Bromo-5-chloro-2-methoxyaniline 95%
4-Bromo-5-chloro-2-methoxyaniline
Precaución: for Research Use Only


Formula: C₇H₇BrClNO
MDL Number: MFCD09878150
CAS Number: 102170-53-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty