Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrimidine-2-carboximidamide acetate 98%

Supplier: AMBEED, INC
Warning

A174153-250MG A174153-5G
77163-674EA 30 USD InStock
77163-674 77163-676
Pyrimidine-2-carboximidamide acetate 98%
Pyrimidine-2-carboximidamide acetate
Precaución: for Research Use Only


Formula: C₇H₁₀N₄O₂
MDL Number: MFCD28369440
CAS Number: 312613-82-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty