Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodotubercidin 98%

Supplier: AMBEED, INC
Warning

A177991-10MG A177991-1MG A177991-25MG A177991-2MG A177991-50MG A177991-5MG
77163-770EA 14 USD InStock
77163-770 77163-772 77163-774 77163-776 77163-778 77163-780
5-Iodotubercidin 98%
5-Iodotubercidin
Precaución: For research use only.


Formula: C₁₁H₁₃IN₀₄
MW: 392.1 g/mol
MDL Number: MFCD00055131
CAS Number: 24386-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty