Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

I-BRD9 98%

Supplier: AMBEED, INC
Danger

A178532-10MG A178532-1MG A178532-50MG A178532-5MG
77191-620EA 46 USD InStock
77191-620 77191-622 77191-624 77191-626
I-BRD9 98%
I-BRD9
Precaución: for Research Use Only


Formula: C₂₂H₂₂F₃N₃O₃S₂
MDL Number: MFCD28952791
CAS Number: 1714146-59-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty