Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-5-fluorobenzyl chloride 98%

Supplier: AMBEED, INC
Danger

A182173-100MG A182173-1G A182173-250MG
77163-906EA 5 USD InStock
77163-906 77163-908 77163-910
2-Cyano-5-fluorobenzyl chloride 98%
2-Cyano-5-fluorobenzyl chloride
Precaución: for Research Use Only


Formula: C₈H₅ClFN
MW: 169.583 g/mol
MDL Number: MFCD18394047
CAS Number: 1261742-22-6
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty