Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Benzo[d]imidazol-2-yl)-4,5-dihydrooxazole 97%

Supplier: AMBEED, INC
Warning

A182707-100MG A182707-1G A182707-250MG
77191-734EA 30 USD InStock
77191-734 77191-736 77191-738
2-(1H-Benzo[d]imidazol-2-yl)-4,5-dihydrooxazole 97%
2-(1H-Benzo[d]imidazol-2-yl)-4,5-dihydrooxazole
Precaución: for Research Use Only


Formula: C₁₀H₉N₃O
MDL Number: MFCD00460180
CAS Number: 14483-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty