Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methylcyclocytidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A183299-100MG A183299-250MG
77163-938EA 6 USD InStock
77163-938 77163-940
5-Methylcyclocytidine hydrochloride 95%
5-Methylcyclocytidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₄ClN₃O₄
MDL Number: MFCD12911590
CAS Number: 51391-96-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty