Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-prop-2-ynylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A184420-100MG A184420-1G A184420-250MG A184420-5G
77163-990EA 17 USD InStock
77163-990 77163-992 77163-994 77163-996
1-Methyl-prop-2-ynylamine hydrochloride 95%
1-Methyl-prop-2-ynylamine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₇N·ClH
MW: 105.566 g/mol
MDL Number: MFCD08752577
CAS Number: 42105-26-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty