Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Cyanomethyl)tetrahydropyran 97%

Supplier: AMBEED, INC
Warning

A184761-10G A184761-1G A184761-250MG A184761-5G
77164-008EA 121 USD InStock
77164-008 77164-010 77164-012 77164-014
4-(Cyanomethyl)tetrahydropyran 97%
4-(Cyanomethyl)tetrahydropyran
Precaución: for Research Use Only


Formula: C₇H₁₁NO
MDL Number: MFCD04973733
CAS Number: 850429-50-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty