Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-ethoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A184960-10G A184960-1G A184960-5G
77191-796EA 45 USD InStock
77191-796 77191-798 77191-800
5-Chloro-2-ethoxyphenylboronic acid 98%
5-Chloro-2-ethoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₀BClO₃
MW: 200.43 g/mol
MDL Number: MFCD03427055
CAS Number: 352534-86-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty