Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Methoxy-2,2,2-trifluoroacetophenone 97%

Supplier: AMBEED, INC
Warning

A187094-10G A187094-1G A187094-25G A187094-5G
77164-100EA 53 USD InStock
77164-100 77164-102 77164-104 77164-106
4'-Methoxy-2,2,2-trifluoroacetophenone 97%
4'-Methoxy-2,2,2-trifluoroacetophenone
Precaución: For research use only.


Formula: C₉H₇F₃O₂
MDL Number: MFCD00018061
CAS Number: 711-38-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty