Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Aminobutyric acid 98%

Supplier: AMBEED, INC
Warning

A189069-1G A189069-250MG A189069-25G A189069-5G A189069-100G
77191-906EA 11 USD InStock
77191-906 77191-908 77191-910 77191-912 77746-211
(S)-3-Aminobutyric acid 98%
(S)-3-Aminobutyric acid
Precaución: for Research Use Only


Formula: C₄H₉NO₂
MW: 103.1200 g/mol
MDL Number: MFCD00270347
CAS Number: 3775-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty