Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-nitro-N-phenylbenzamide 97%

Supplier: AMBEED, INC
Warning

A189155-10G A189155-1G A189155-5G
77191-914EA 38 USD InStock
77191-914 77191-916 77191-918 77191-920
2-Fluoro-6-nitro-N-phenylbenzamide 97%
2-Fluoro-6-nitro-N-phenylbenzamide
Precaución: for Research Use Only


Formula: C₁₃H₉FN₂O₃
MDL Number: MFCD16875530
CAS Number: 870281-83-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty