Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-fluoro-4-methylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A189808-100MG A189808-1G A189808-250MG
77164-216EA 15 USD InStock
77164-216 77164-218 77164-220
2-Amino-5-fluoro-4-methylbenzoic acid 97%
2-Amino-5-fluoro-4-methylbenzoic acid


Formula: C₈H₈FNO₂
MDL Number: MFCD18300404
CAS Number: 103877-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty