Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrazolo[4,3-b]pyridine 98%

Supplier: AMBEED, INC
Warning

A190221-100MG A190221-1G A190221-250MG A190221-5G
77164-248EA 6 USD InStock
77164-248 77164-250 77164-252 77164-254
1H-Pyrazolo[4,3-b]pyridine 98%
1H-Pyrazolo[4,3-b]pyridine
Precaución: for Research Use Only


Formula: C₆H₅N₃
MDL Number: MFCD08234560
CAS Number: 272-52-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty