Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloroquinoline 98%

Supplier: AMBEED, INC
Danger

A190365-1G A190365-250MG A190365-5G
77164-260EA 39 USD InStock
77164-260 77164-262 77164-264
4,6-Dichloroquinoline 98%
4,6-Dichloroquinoline


Formula: C₉H₅Cl₂N
MDL Number: MFCD00156141
CAS Number: 4203-18-3
UN: 2923
ADR: 8,6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty