Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A192122-10G A192122-1G A192122-250MG A192122-25G A192122-5G
77191-992EA 24 USD InStock
77191-992 77191-994 77191-996 77191-998 77192-000
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₂₁BN₂O₂
MDL Number: MFCD06659062
CAS Number: 844891-04-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty