Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-4-[(4-methoxybenzyl)oxy]-2-nitrobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A192261-100G A192261-1G A192261-25G
77164-298EA 434 USD InStock
77164-298 77164-300 77164-302
5-Methoxy-4-[(4-methoxybenzyl)oxy]-2-nitrobenzoic acid 97%
5-Methoxy-4-[(4-methoxybenzyl)oxy]-2-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₁₆H₁₅NO₇
MDL Number: MFCD28144964
CAS Number: 1643979-88-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty