Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Amino-1-cyclobutanecarboxylic acid hydrochloride 95%

Supplier: AMBEED, INC
Warning

A193293-100G A193293-1G A193293-25G A193293-5G
77192-030EA 362.99 USD InStock
77192-030 77192-032 77192-034 77192-036
1-Amino-1-cyclobutanecarboxylic acid hydrochloride 95%
1-Amino-1-cyclobutanecarboxylic acid hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₀ClNO₂
MDL Number: MFCD08752285
CAS Number: 98071-16-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty