Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-decyl-3-methylimidazolium hexafluorophosphate 99%

Supplier: AMBEED, INC
Warning

A193468-1G A193468-25G A193468-5G
77192-042EA 6 USD InStock
77192-042 77192-044 77192-046
1-decyl-3-methylimidazolium hexafluorophosphate 99%
1-decyl-3-methylimidazolium hexafluorophosphate
Precaución: for Research Use Only


Formula: C₁₄H₂₇F₆N₂P
MDL Number: MFCD12022377
CAS Number: 362043-46-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty