Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Thieno[3,2-d]pyrimidine-2,4-dione 97%

Supplier: AMBEED, INC
Warning

A194007-100G A194007-10G A194007-1G A194007-250MG A194007-5G
77192-074EA 164 USD InStock
77192-074 77192-076 77192-078 77192-080 77192-082
1H-Thieno[3,2-d]pyrimidine-2,4-dione 97%
1H-Thieno[3,2-d]pyrimidine-2,4-dione
Precaución: for Research Use Only


Formula: C₆H₄N₂O₂S
MDL Number: MFCD09909655
CAS Number: 16233-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty