Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Boc-amino)-3-methyl-1-butanol 95%

Supplier: AMBEED, INC
Warning

A194201-100MG A194201-1G A194201-250MG
77192-084EA 27 USD InStock
77192-084 77192-086 77192-088
3-(Boc-amino)-3-methyl-1-butanol 95%
3-(Boc-amino)-3-methyl-1-butanol
Precaución: for Research Use Only


Formula: C₁₀H₂₁NO₃
MDL Number: MFCD19982636
CAS Number: 167216-22-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty