Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palonosetron hydrochloride 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A195751-100MG A195751-10MG A195751-1G A195751-1MG A195751-50MG A195751-5G
77164-400EA 52 USD InStock
77164-400 77164-402 77164-404 77164-406 77164-408 77683-533
Palonosetron hydrochloride 98%, Ambeed.Inc
Palonosetron hydrochloride
Precaución: For research use only.


Formula: C₁₉H₂₄N₂O
MDL Number: MFCD07370072
CAS Number: 135729-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty