Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(4-(2-Chloro-5-iodobenzyl)phenoxy)tetrahydrofuran 97%

Supplier: AMBEED, INC
Warning

A197552-100G A197552-10G A197552-1G A197552-250MG A197552-25G A197552-5G
77164-478EA 83 USD InStock
77164-478 77164-480 77164-482 77164-484 77164-486 77164-488
(S)-3-(4-(2-Chloro-5-iodobenzyl)phenoxy)tetrahydrofuran 97%
(S)-3-(4-(2-Chloro-5-iodobenzyl)phenoxy)tetrahydrofuran
Precaución: for Research Use Only


Formula: C₁₇H₁₆ClIO₂
MDL Number: MFCD28098707
CAS Number: 915095-94-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty