Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 4-bromobenzenesulfinate 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A198284-5G A198284-100MG A198284-250MG
77164-502EA 254 USD InStock
77164-502 77683-545 77683-546
Sodium 4-bromobenzenesulfinate 95%, Ambeed.Inc
Sodium 4-bromobenzenesulphinate
Precaución: For research use only.


Formula: C₆H₄BrNaO₂S
MW: 243.0535 g/mol
MDL Number: MFCD09065037
CAS Number: 34176-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty