Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Adamantanedimethanol 97%

Supplier: AMBEED, INC
Warning

A198727-100MG A198727-1G A198727-250MG
77192-216EA 14 USD InStock
77192-216 77192-218 77192-220
1,3-Adamantanedimethanol 97%
1,3-Adamantanedimethanol
Precaución: for Research Use Only


Formula: C₁₂H₂₀O₂
MDL Number: MFCD00154014
CAS Number: 17071-62-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty