Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CYC116 98+%

Supplier: AMBEED, INC
Warning

A199103-10MG A199103-1MG A199103-50MG A199103-5MG
77192-228EA 62 USD InStock
77192-228 77192-230 77192-232 77192-234
CYC116 98+%
CYC116
Precaución: for Research Use Only


Formula: C₁₈H₂₀N₆OS
MW: 368.5 g/mol
MDL Number: MFCD14155805
CAS Number: 693228-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty