Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Hydroxyoctanoic acid 97%

Supplier: AMBEED, INC
Warning

A200175-1G A200175-250MG A200175-5G A200175-100MG
77192-254EA 21 USD InStock
77192-254 77192-256 77567-884 77746-250
8-Hydroxyoctanoic acid 97%
8-Hydroxyoctanoic acid
Precaución: For research use only.


Formula: C₈H₁₆O₃
MW: 160.21 g/mol
MDL Number: MFCD00792446
CAS Number: 764-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty