Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tavaborole 98%

Supplier: AMBEED, INC
Warning

A200444-100MG A200444-10MG A200444-1MG A200444-250MG A200444-50MG A200444-5G A200444-1G
77164-560EA 10 USD InStock
77164-560 77164-562 77164-564 77164-566 77164-568 77164-570 77567-890
Tavaborole 98%
Tavaborole
Precaución: For research use only.


Formula: C₇H₆BFO₂
MDL Number: MFCD10699483
CAS Number: 174671-46-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty