Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A200448-100G A200448-1G A200448-5G A200448-10G
77164-572EA 155 USD InStock
77164-572 77164-574 77164-576 77683-559
Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%, Ambeed.Inc
Ethyl-trans-4-hydroxycyclohexanecarboxylate
Precaución: For research use only.


Formula: C₉H₁₆O₃
MW: 172.22 g/mol
MDL Number: MFCD16295187
CAS Number: 3618-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty