Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Boc-aminomethyl)cyclobutanone 97%

Supplier: AMBEED, INC
Warning

A202120-100MG A202120-1G A202120-250MG A202120-5G
77164-620EA 26 USD InStock
77164-620 77164-622 77164-624 77164-626
3-(Boc-aminomethyl)cyclobutanone 97%
3-(Boc-aminomethyl)cyclobutanone
Precaución: for Research Use Only


Formula: C₁₀H₁₇NO₃
MDL Number: MFCD11499004
CAS Number: 130369-09-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty