Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2R)-trans-2-(Boc-amino)cyclohexanol hydrochloride 97%

Supplier: AMBEED, INC
Warning

A203345-1G A203345-250MG A203345-5G
77164-660EA 12 USD InStock
77164-660 77164-662 77164-664
(1R,2R)-trans-2-(Boc-amino)cyclohexanol hydrochloride 97%
(1R,2R)-trans-2-(Boc-amino)cyclohexanol hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₃
MDL Number: MFCD11656036
CAS Number: 155975-19-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty