Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl (3-fluoro-4-thiomorpholinophenyl)carbamate 97%

Supplier: AMBEED, INC
Warning

A203808-100G A203808-1G A203808-250MG A203808-25G A203808-5G
77192-392EA 1048 USD InStock
77192-392 77192-394 77192-396 77192-398 77192-400
Benzyl (3-fluoro-4-thiomorpholinophenyl)carbamate 97%
Benzyl (3-fluoro-4-thiomorpholinophenyl)carbamate
Precaución: for Research Use Only


Formula: C₁₈H₁₉FN₂O₂S
MDL Number: MFCD28016162
CAS Number: 168828-71-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty