Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2,4-dichloropyrimidine 98%

Supplier: AMBEED, INC
Warning

A204706-100G A204706-10G A204706-1G A204706-25G A204706-5G
77164-708EA 287.99 USD InStock
77164-708 77164-710 77164-712 77164-714 77164-716
5-Amino-2,4-dichloropyrimidine 98%
5-Amino-2,4-dichloropyrimidine
Precaución: for Research Use Only


Formula: C₄H₃Cl₂N₃
MDL Number: MFCD05662684
CAS Number: 5177-27-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty