Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodoisoquinolin-1-amine 96%

Supplier: AMBEED, INC
Warning

A206567-100MG A206567-1G
77164-792EA 17 USD InStock
77164-792 77164-794
4-Iodoisoquinolin-1-amine 96%
4-Iodoisoquinolin-1-amine
Precaución: for Research Use Only


Formula: C₉H₇IN₂
MDL Number: MFCD03695827
CAS Number: 55270-28-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty