Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrrolidine-2-carbonitrile 4-methylbenzenesulfonate 98%

Supplier: AMBEED, INC
Warning

A206690-1G A206690-250MG A206690-25G A206690-5G
77164-798EA 55 USD InStock
77164-798 77164-800 77164-802 77164-804
Pyrrolidine-2-carbonitrile 4-methylbenzenesulfonate 98%
Pyrrolidine-2-carbonitrile 4-methylbenzenesulfonate
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂O₃S
MW: 268.33 g/mol
MDL Number: MFCD28400995
CAS Number: 1666113-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty