Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Iodo-m-toluidine 98%

Supplier: AMBEED, INC
Warning

A207369-100G A207369-10G A207369-1G A207369-25G A207369-500G A207369-5G
77192-514EA 119 USD InStock
77192-514 77192-516 77192-518 77192-520 77192-522 77192-524 77192-526
6-Iodo-m-toluidine 98%
6-Iodo-m-toluidine
Precaución: for Research Use Only


Formula: C₇H₈IN
MDL Number: MFCD00833395
CAS Number: 13194-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty