Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis(2-nitrophenoxy)ethane 98%

Supplier: AMBEED, INC
Warning

A207721-100G A207721-1G A207721-25G A207721-5G
77192-536EA 221 USD InStock
77192-536 77192-538 77192-540 77192-542
1,2-Bis(2-nitrophenoxy)ethane 98%
1,2-Bis(2-nitrophenoxy)ethane
Precaución: for Research Use Only


Formula: C₁₄H₁₂N₂O₆
MDL Number: MFCD00024226
CAS Number: 51661-19-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty