Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl isobutyrylacetate 97%

Supplier: AMBEED, INC
Danger

A207849-10G A207849-1G A207849-25G A207849-5G
77192-544EA 13 USD InStock
77192-544 77192-546 77192-548 77192-550
Ethyl isobutyrylacetate 97%
Ethyl isobutyrylacetate
Precaución: for Research Use Only


Formula: C₈H₁₄O₃
MDL Number: MFCD00009198
CAS Number: 7152-15-0
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty