Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-N-(4-Hydroxy-3-methoxybenzyl)-8-methylnon-6-enamide 95%

Supplier: AMBEED, INC
Warning

A208947-10MG A208947-1MG A208947-250MG A208947-50MG
77192-566EA 21 USD InStock
77192-566 77192-568 77192-570 77192-572
(Z)-N-(4-Hydroxy-3-methoxybenzyl)-8-methylnon-6-enamide 95%
(Z)-N-(4-Hydroxy-3-methoxybenzyl)-8-methylnon-6-enamide
Precaución: for Research Use Only


Formula: C₁₈H₂₇NO₃
MDL Number: MFCD00209942
CAS Number: 25775-90-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty