Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3,4-Dichlorophenyl)pyrrolidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A209576-250MG A209576-50MG
77164-900EA 176 USD InStock
77164-900 77164-902
3-(3,4-Dichlorophenyl)pyrrolidine hydrochloride 95%
3-(3,4-Dichlorophenyl)pyrrolidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₂Cl₃N
MDL Number: MFCD28369479
CAS Number: 1092108-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty