Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-(4'-Bromo-4-biphenylyl)carbazole 98%

Supplier: AMBEED, INC
Warning

A209873-1G A209873-25G A209873-5G
77192-584EA 12 USD InStock
77192-584 77192-586 77192-588
9-(4'-Bromo-4-biphenylyl)carbazole 98%
9-(4'-Bromo-4-biphenylyl)carbazole
Precaución: for Research Use Only


Formula: C₂₄H₁₆BrN
MDL Number: MFCD27644692
CAS Number: 212385-73-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty