Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Choline-p-toluenesulfonate 95%

Supplier: AMBEED, INC
Warning

A210585-100G A210585-10G A210585-25G A210585-5G
77192-624EA 158 USD InStock
77192-624 77192-626 77192-628 77192-630
Choline-p-toluenesulfonate 95%
Choline p-toluenesulphonate
Precaución: for Research Use Only


Formula: C₅H₁₄NO
MDL Number: MFCD00079051
CAS Number: 55357-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty