Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

15-Hydroxypentadecanoic acid 98%

Supplier: AMBEED, INC
Warning

A214041-1G A214041-250MG A214041-25G A214041-5G
77192-730EA 20 USD InStock
77192-730 77192-732 77192-734 77192-736
15-Hydroxypentadecanoic acid 98%
15-Hydroxypentadecanoic acid
Precaución: for Research Use Only


Formula: C₁₅H₃₀O₃
MW: 258.40 g/mol
MDL Number: MFCD00039535
CAS Number: 4617-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty