Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyano-5-pyridineboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A214344-1G A214344-250MG A214344-25G A214344-5G
77165-056EA 16 USD InStock
77165-056 77165-058 77165-060 77165-062
2-Cyano-5-pyridineboronic acid pinacol ester 97%
2-Cyano-5-pyridineboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₅BN₂O₂
MW: 230.07
MDL Number: MFCD06657825
CAS Number: 741709-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty