Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,5-dimethylthiazole 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A214514-100MG A214514-1G
77165-064EA 30 USD InStock
77165-064 77683-609
4-Bromo-2,5-dimethylthiazole 95%, Ambeed.Inc
4-Bromo-2,5-dimethylthiazole
Precaución: For research use only.


Formula: C₅H₆BrNS
MDL Number: MFCD20491436
CAS Number: 90672-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty