Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,1,3,5,7,7,7-Octamethyltetrasiloxane 98%

Supplier: AMBEED, INC
Warning

A216058-100G A216058-500G
77165-088EA 35 USD InStock
77165-088 77165-090 77165-092
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane 98%
1,1,1,3,5,7,7,7-Octamethyltetrasiloxane
Precaución: for Research Use Only


Formula: C₈H₂₆O₃Si₄
MDL Number: MFCD00053664
CAS Number: 16066-09-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty